siRNA Details
Gene Symbol: Helq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081107.1 | NP_001074576.1 | 1, 2 | 315 | ||
GenBank mRNA | AK040335.1 | - | 1, 2 | 299 | ||
AK050769.1 | - | 1, 2 | 292 | |||
AK052427.1 | - | 1, 2 | 275 | |||
BC082601.1 | - | 1, 2 | 279 | 30662103 | ||
BC109169.2 | - | 1, 2 | 183 | |||
BC109170.2 | - | 1, 2 | 188 | 40048258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
