siRNA Details
Gene Symbol: Plekhh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181073.3 | NP_851418.2 | 27, 28 | 4085 | ||
GenBank mRNA | AK052692.1 | - | 14, 15 | 1760 | ||
AK122464.1 | - | 27, 28 | 4054 | |||
AK143387.1 | - | 27, 28 | 3902 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
