siRNA Details
Gene Symbol: PRKD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079880.1 | NP_001073349.1 | 9 | 1689 | ||
NM_001079881.1 | NP_001073350.1 | 9 | 1553 | |||
NM_001079882.1 | NP_001073351.1 | 7 | 1290 | |||
NM_016457.4 | NP_057541.2 | 8 | 1968 | |||
XM_005258716.2 | XP_005258773.2 | 7 | 946 | |||
GenBank mRNA | AF309082.1 | - | 8 | 1250 | ||
AK074673.1 | - | 7 | 929 | |||
AK075266.1 | - | 7 | 1039 | |||
AK092896.1 | - | 8 | 1340 | |||
AK095884.1 | - | 9 | 1232 | |||
AK300339.1 | - | 9 | 1503 | |||
AK303629.1 | - | 7 | 944 | |||
AL050147.1 | - | 2 | 205 | |||
BC025307.1 | - | 4 | 539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
