siRNA Details
Gene Symbol: Anks1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128086.2 | NP_001121558.2 | 16 | 2706 | ||
NM_001177396.1 | NP_001170867.1 | 2 | 537 | |||
NM_001177397.1 | NP_001170868.1 | 2 | 152 | |||
NM_001177398.1 | NP_001170869.1 | 2 | 537 | |||
NM_181398.3 | NP_852063.1 | 2 | 537 | |||
GenBank mRNA | AK032211.1 | - | 2 | 539 | ||
AK034154.1 | - | 3 | 172 | |||
AK049899.1 | - | 2 | 482 | |||
AK132228.1 | - | 2 | 504 | |||
AK163750.1 | - | 2 | 511 | |||
BC098373.1 | - | 2 | 152 |
Gene Symbol: Ipw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019361.1 | - | 481 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001271371, XM_006241248, XM_006241249, XM_008765239, XM_008765241, and others.
Gene Ontology Categories:
