siRNA Details
Gene Symbol: Kiss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF472576.1 | - | 2 | 361 | ||
BC117046.1 | - | 2 | 346 | 40126365 | ||
BC119348.1 | - | 2 | 346 | 8734097 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
