siRNA Details
Gene Symbol: Sirpb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB109404.1 | - | 946 | |||
AB112023.1 | - | 872 | ||||
AB112025.1 | - | 3 | 452 | |||
AK036935.1 | - | 864 | ||||
AK040982.1 | - | 922 | ||||
AK041165.1 | - | 926 | ||||
AK054545.1 | - | 882 | ||||
AK170671.1 | - | 872 | ||||
BC145417.1 | - | 881 | 9053975 | |||
BC145418.1 | - | 882 | 9053976 |
Gene Symbol: LOC100038947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173459.2 | NP_001166930.1 | 4 | 921 | ||
GenBank mRNA | AK155731.1 | - | 922 | |||
DQ055453.1 | - | 4 | 870 |
Gene Symbol: Sirpb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173460.1 | NP_001166931.1 | 4 | 962 | ||
GenBank mRNA | AK154879.1 | - | 4 | 963 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
