siRNA Details
Gene Symbol: A530064D06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113556.1 | NP_001107028.1 | 2 | 376 | ||
NM_178796.6 | NP_848911.2 | 2 | 376 | |||
GenBank mRNA | AB372005.1 | - | 2 | 153 | ||
AK080114.1 | - | 2 | 337 | |||
AK089332.1 | - | 2 | 337 | |||
AK170607.1 | - | 2 | 395 | |||
AK171433.1 | - | 337 | ||||
BC139185.1 | - | 2 | 211 | 8862207 | ||
BC139186.1 | - | 2 | 211 | 8862208 | ||
BC145572.1 | - | 2 | 211 | 9054138 | ||
EU003111.1 | - | 2 | 107 | |||
EU003112.1 | - | 2 | 107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
