siRNA Details
Gene Symbol: ALPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001632.4 | NP_001623.3 | 11 | 2812 | ||
GenBank mRNA | AF217992.1 | - | 1 | 969 | ||
BC009647.1 | - | 11 | 2609 | 3460735 | ||
BC068501.1 | - | 11 | 2595 | 30349481 | ||
BC094743.1 | - | 11 | 2675 | 30330997 | ||
M12551.1 | - | 11 | 2552 | |||
M14169.1 | - | 10 | 2492 | |||
M14170.1 | - | 11 | 2556 |
Gene Symbol: GPALPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011535144.2 | XP_011533446.1 | 9 | 3027 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
