siRNA Details
Gene Symbol: AMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287215.1 | NP_001274144.1 | 2 | 273 | ||
NM_001634.5 | NP_001625.2 | 2 | 510 | |||
GenBank mRNA | AK304351.1 | - | 2 | 273 | ||
AK316471.1 | - | 2 | 142 | |||
AU134914.1 | - | 2 | 510 | |||
BC000171.2 | - | 2 | 283 | 2900587 | ||
BX640599.1 | - | 2 | 510 | |||
M21154.1 | - | 2 | 436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
