siRNA Details
Gene Symbol: Anks1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128086.2 | NP_001121558.2 | 18 | 2963 | ||
NM_001177396.1 | NP_001170867.1 | 4 | 794 | |||
NM_001177397.1 | NP_001170868.1 | 4 | 409 | |||
NM_001177398.1 | NP_001170869.1 | 4 | 794 | |||
NM_181398.3 | NP_852063.1 | 4 | 794 | |||
GenBank mRNA | AK032061.1 | - | 2 | 65 | ||
AK032211.1 | - | 4 | 796 | |||
AK034154.1 | - | 5 | 429 | |||
AK038609.1 | - | 2 | 293 | |||
AK049899.1 | - | 4 | 739 | |||
AK132228.1 | - | 4 | 761 | |||
AK163750.1 | - | 4 | 768 | |||
BC098373.1 | - | 4 | 409 |
Gene Symbol: Ipw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019361.1 | - | 738 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
