siRNA Details
Gene Symbol: Zbed4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181412.3 | NP_852077.1 | 3 | 734 | ||
GenBank mRNA | AK122335.1 | - | 3 | 704 | ||
BC052174.1 | - | 2 | 637 | 6518433 | ||
BC092524.1 | - | 1 | 358 |
