siRNA Details
Gene Symbol: Slc8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112798.2 | NP_001106269.1 | 9 | 16424 | ||
NM_001286684.1 | NP_001273613.1 | 12 | 13698 | |||
NM_011406.3 | NP_035536.2 | 11 | 16460 | |||
GenBank mRNA | AK078052.1 | - | 1 | 1485 | ||
BC060264.1 | - | 1 | 441 | |||
BC094585.1 | - | 1 | 590 | |||
BC096032.1 | - | 2 | 289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
