siRNA Details
Gene Symbol: Helq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081107.1 | NP_001074576.1 | 2 | 794 | ||
GenBank mRNA | AK040335.1 | - | 2 | 778 | ||
AK050769.1 | - | 2 | 771 | |||
AK052427.1 | - | 2 | 754 | |||
BC082601.1 | - | 2 | 758 | 30662103 | ||
BC109169.2 | - | 2 | 662 | |||
BC109170.2 | - | 2 | 667 | 40048258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
