siRNA Details
Gene Symbol: Hba-x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010405.4 | NP_034535.1 | 1, 2 | 156 | ||
GenBank mRNA | AK077590.1 | - | 1, 2 | 155 | ||
BC051988.1 | - | 1, 2 | 126 | 5711898 | ||
BY111754.1 | - | 1, 2 | 157 | |||
M26898.1 | - | 1, 2 | 114 |
