siRNA Details
Gene Symbol: Erc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053204.2 | NP_444434.2 | 21 | 8652 | ||
GenBank mRNA | AK019888.1 | - | 1 | 840 | ||
BC002272.1 | - | 1 | 231 | |||
BC057059.1 | - | 1 | 2164 | |||
BC079639.1 | - | 1 | 883 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
