siRNA Details
Gene Symbol: ZAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016653.2 | NP_057737.2 | 16, 17 | 1567 | ||
XM_005246640.2 | XP_005246697.1 | 16, 17 | 1613 | |||
GenBank mRNA | AB030034.1 | - | 16, 17 | 1542 | ||
AB049733.1 | - | 15, 16 | 1367 | |||
AB208974.1 | - | 16, 17 | 1503 | |||
AF238255.1 | - | 15, 16 | 1367 | |||
AF251441.1 | - | 16, 17 | 1445 | |||
AF480461.1 | - | 16, 17 | 1562 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
