siRNA Details
Gene Symbol: Zfp65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145622.2 | NP_663597.1 | 4 | 2282 | ||
GenBank mRNA | AK135182.1 | - | 1 | 787 | ||
BC016248.1 | - | 3 | 2191 | 4527362 |
Gene Symbol: 1700008H02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005767.1 | - | 1 | 298 |
Gene Symbol: 4931432E15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016500.1 | - | 1 | 1565 |
Gene Symbol: 4933411E06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016768.1 | - | 1 | 241 |
Gene Symbol: 6530403H02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032656.1 | - | 14 | 2172 |
Gene Symbol: A130077B15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038082.1 | - | 3 | 1638 |
Gene Symbol: Bora | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC052183.1 | - | 10 | 1760 | 6336038 |
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK166883.1 | - | 3 | 2851 |
Gene Symbol: Gatad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK081359.1 | - | 2306 |
Gene Symbol: Gm15614 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136552.1 | - | 4 | 1164 |
Gene Symbol: Gm16041 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK044237.1 | - | 3 | 697 |
Gene Symbol: Gm3871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK148631.1 | - | 1 | 1550 |
Gene Symbol: Gm39182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038453.1 | - | 382 |
Gene Symbol: Heatr5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008221.1 | - | 242 | |||
BC019508.1 | - | 246 |
Gene Symbol: Hsd3b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC040397.1 | - | 2794 | 4222081 |
Gene Symbol: Jakmip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK048388.1 | - | 1 | 1664 |
Gene Symbol: Macrod2os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031201.1 | - | 4 | 1331 |
Gene Symbol: Ranbp3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132406.1 | - | 2953 |
Gene Symbol: Skint8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076260.1 | - | 1 | 1210 |
Gene Symbol: Sorbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK166943.1 | - | 4 | 2366 |
Gene Symbol: Syne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079686.1 | NP_001073154.1 | 55 | 9230 | ||
NM_022027.2 | NP_071310.2 | 17 | 3184 | |||
GenBank mRNA | AF281869.1 | - | 17 | 3100 | ||
AF281870.1 | - | 36 | 6141 | |||
AK036828.1 | - | 18 | 3240 | |||
BC041779.1 | - | 16 | 2781 | |||
BC054456.1 | - | 17 | 3184 | |||
BC110426.1 | - | 25 | 4433 | |||
BC110428.1 | - | 17 | 3164 | |||
BC111034.1 | - | 6 | 980 |
Gene Symbol: Zrsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038641.1 | - | 2713 |

More Information
Other Species Targeted

Gene Ontology Categories:
