siRNA Details
Gene Symbol: Krtap9-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029351.2 | NP_083627.2 | 1 | 464 | ||
GenBank mRNA | AK009730.1 | - | 1 | 466 | ||
AK131927.1 | - | 1 | 465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
