siRNA Details
Gene Symbol: Neurog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009719.6 | NP_033849.3 | 2 | 469 | ||
GenBank mRNA | BC104326.2 | - | 1 | 518 | 40045043 | |
BC104327.2 | - | 1 | 518 | 40045044 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
