siRNA Details
Gene Symbol: Hist2h2aa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013549.2 | NP_038577.1 | 1 | 452 | ||
GenBank mRNA | AK002725.1 | - | 1 | 412 | ||
AK006728.1 | - | 1 | 411 | |||
BC010564.1 | - | 1 | 395 | 3582122 | ||
BC053966.1 | - | 1 | 404 | |||
BC062255.1 | - | 1 | 395 | 1448126 | ||
BC064002.1 | - | 394 | ||||
BC089519.1 | - | 1 | 452 | 6771160 | ||
BC147778.1 | - | 1 | 449 | 9087498 | ||
BC147781.1 | - | 1 | 449 | 9087501 | ||
Z30940.1 | - | 1 | 390 |
Gene Symbol: Hist1h2al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077055.1 | - | 226 |
Gene Symbol: Hist2h2aa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178212.3 | NP_835584.1 | 1 | 492 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
