siRNA Details
Gene Symbol: Defb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF155882.2 | - | 2 | 216 | ||
BC099882.1 | - | 2 | 236 | 40040625 | ||
BC099883.1 | - | 2 | 236 | 40040627 | ||
BC100674.1 | - | 2 | 236 | 40040624 | ||
BC100675.1 | - | 2 | 236 | 40040630 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
