siRNA Details
Gene Symbol: Nudt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021431.2 | NP_067406.2 | 1 | 766 | ||
GenBank mRNA | AB041576.1 | - | 1 | 512 | ||
AK036077.1 | - | 1 | 97 | |||
AK053310.1 | - | 1 | 546 | |||
AY152853.1 | - | 1 | 512 | |||
BC071274.1 | - | 1 | 570 | 6331160 |
Gene Symbol: Nudt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031664.1 | NP_001026834.1 | 1 | 518 | ||
GenBank mRNA | BC055771.1 | - | 1 | 475 | 6404994 | |
CX209897.1 | - | 1 | 518 |

More Information
Other Species Targeted

Gene Ontology Categories:
