siRNA Details
Gene Symbol: Mmp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080453.2 | NP_536701.1 | 7 | 1135 | ||
GenBank mRNA | AK151100.1 | - | 7 | 1198 | ||
AY065653.1 | - | 7 | 1165 | |||
AY071828.1 | - | 7 | 1135 | |||
BC145048.1 | - | 7 | 1120 | 9053596 | ||
BC145049.1 | - | 7 | 1090 | 9053597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
