siRNA Details
Gene Symbol: Obox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145708.2 | NP_663754.2 | 2 | 291 | ||
GenBank mRNA | BC094912.1 | - | 2 | 171 | ||
CA556780.1 | - | 2 | 191 | |||
DV663188.1 | - | 2 | 290 |
Gene Symbol: Obox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145709.2 | NP_663755.2 | 2 | 249 | ||
GenBank mRNA | BC052664.1 | - | 2 | 253 | 30053785 | |
CJ052752.1 | - | 2 | 251 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
