siRNA Details
Gene Symbol: Suox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173733.3 | NP_776094.2 | 4 | 2174 | ||
GenBank mRNA | AK149422.1 | - | 4 | 2162 | ||
AK154608.1 | - | 4 | 2162 | |||
BC027197.1 | - | 3 | 2129 | 4160277 |

More Information
Other Species Targeted

Gene Ontology Categories:
