siRNA Details
Gene Symbol: Mex3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175366.3 | NP_780575.2 | 2 | 2763 | ||
GenBank mRNA | AK077009.1 | - | 1 | 1370 | ||
AK088538.1 | - | 1 | 211 | |||
BC067199.1 | - | 1 | 1729 | |||
BC108401.1 | - | 1 | 1973 |

More Information
Other Species Targeted

Gene Ontology Categories:
