siRNA Details
Gene Symbol: Hist2h2ac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175662.2 | NP_783593.1 | 1 | 401 | ||
GenBank mRNA | BC117094.1 | - | 1 | 401 | 40126413 | |
BC117096.1 | - | 1 | 401 | 40126415 |
Gene Symbol: Hist1h2aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175658.2 | NP_783589.1 | 1 | 365 | ||
GenBank mRNA | BC107354.1 | - | 1 | 365 | 40055855 | |
BC107355.1 | - | 1 | 365 | 40055860 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
