siRNA Details
Gene Symbol: B430306N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177083.4 | NP_796057.1 | 2 | 346 | ||
GenBank mRNA | AK036709.1 | - | 2 | 347 | ||
AK046672.1 | - | 347 | ||||
AK133242.1 | - | 2 | 1207 | |||
AK155881.1 | - | 2 | 362 | |||
AK157919.1 | - | 2 | 85 | |||
AK170621.1 | - | 2 | 368 | |||
AK171230.1 | - | 2 | 345 | |||
AK171587.1 | - | 2 | 347 | |||
AK171603.1 | - | 2 | 345 | |||
AK171605.1 | - | 2 | 362 | |||
AY522648.1 | - | 2 | 347 | |||
DQ087179.1 | - | 2 | 99 | |||
DQ087180.1 | - | 2 | 99 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
