siRNA Details
Gene Symbol: Sybu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043739.1 | - | 2 | 508 | ||
AK047449.1 | - | 2 | 498 | |||
AK049811.1 | - | 2 | 442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
