siRNA Details
Gene Symbol: Crb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177638.4 | NP_808306.1 | 4 | 403 | ||
GenBank mRNA | AK077528.1 | - | 4 | 335 | ||
AK167279.1 | - | 4 | 333 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
