siRNA Details
Gene Symbol: Klrb1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153094.2 | NP_694734.1 | 6 | 808 | ||
GenBank mRNA | AK041601.1 | - | 6 | 809 | ||
AK154278.1 | - | 6 | 795 | |||
AK170023.1 | - | 6 | 785 | |||
BC107177.1 | - | 5 | 635 | |||
BC107178.1 | - | 5 | 635 | |||
BC127141.1 | - | 6 | 710 | |||
BC127142.1 | - | 6 | 785 | 40050668 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
