siRNA Details
Gene Symbol: B430306N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177083.4 | NP_796057.1 | 2 | 442 | ||
GenBank mRNA | AK036709.1 | - | 2 | 443 | ||
AK046672.1 | - | 443 | ||||
AK133242.1 | - | 2 | 1303 | |||
AK155881.1 | - | 2 | 458 | |||
AK157919.1 | - | 2 | 181 | |||
AK170621.1 | - | 2 | 464 | |||
AK171230.1 | - | 2 | 441 | |||
AK171587.1 | - | 2 | 443 | |||
AK171603.1 | - | 2 | 441 | |||
AK171605.1 | - | 2 | 458 | |||
AY522648.1 | - | 2 | 443 | |||
DQ087179.1 | - | 2 | 195 | |||
DQ087180.1 | - | 2 | 195 |
Gene Symbol: 9830107B12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177896.1 | NP_001171367.1 | 2 | 475 | ||
NM_177824.4 | NP_808492.1 | 2 | 475 | |||
GenBank mRNA | AK079398.1 | - | 2 | 436 | ||
EF646801.1 | - | 176 |
Gene Symbol: A530064D06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113556.1 | NP_001107028.1 | 2 | 445 | ||
NM_178796.6 | NP_848911.2 | 2 | 445 | |||
GenBank mRNA | AB372005.1 | - | 2 | 222 | ||
AK080114.1 | - | 2 | 406 | |||
AK089248.1 | - | 2 | 459 | |||
AK089332.1 | - | 2 | 406 | |||
AK170607.1 | - | 2 | 464 | |||
AK171433.1 | - | 406 | ||||
BC139185.1 | - | 2 | 280 | 8862207 | ||
BC139186.1 | - | 2 | 280 | 8862208 | ||
BC145572.1 | - | 2 | 280 | 9054138 | ||
EU003111.1 | - | 2 | 176 | |||
EU003112.1 | - | 2 | 176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
