siRNA Details
Gene Symbol: Aph1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177583.4 | NP_808251.1 | 6 | 2296 | ||
GenBank mRNA | AK028521.1 | - | 7 | 2555 | ||
AK049828.1 | - | 6 | 2297 | |||
AK154847.1 | - | 6 | 2364 | |||
AK162585.1 | - | 7 | 2531 | |||
BC079659.1 | - | 6 | 2328 | 6853991 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
