siRNA Details
Gene Symbol: Fmn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285458.1 | NP_001272387.1 | 17 | 6085 | ||
NM_001285459.1 | NP_001272388.1 | 17 | 5709 | |||
NM_001287817.1 | NP_001274746.1 | 5 | 3589 | |||
NM_001287818.1 | NP_001274747.1 | 6 | 2329 | |||
NM_010230.2 | NP_034360.2 | 20 | 6487 | |||
GenBank mRNA | AK148751.1 | - | 1 | 1413 |
Gene Symbol: BC022713 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052913.1 | - | 1 | 2710 | ||
BC022713.1 | - | 1 | 696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
