siRNA Details
Gene Symbol: Defa-rs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007844.2 | NP_031870.2 | 1, 2 | 219 | ||
GenBank mRNA | AK008584.1 | - | 1, 2 | 220 | ||
BC117042.1 | - | 1, 2 | 194 | 40126361 | ||
BC117044.1 | - | 1, 2 | 194 | 40126363 | ||
BC145556.1 | - | 1, 2 | 224 | 9054120 | ||
BC171966.1 | - | 1, 2 | 224 | 9054252 | ||
EU760894.1 | - | 15 | ||||
M33226.1 | - | 1, 2 | 199 |
Gene Symbol: AY761184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EU760895.1 | - | 15 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
