siRNA Details
Gene Symbol: Cma2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024714.2 | NP_001019885.1 | 2 | 169 | ||
GenBank mRNA | AF361939.1 | - | 2 | 169 | ||
BC139236.1 | - | 2 | 164 | 8862258 | ||
BC139237.1 | - | 2 | 164 | 8862259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
