siRNA Details
Gene Symbol: IFIT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010987.2 | NP_001010987.1 | 2 | 794 | ||
GenBank mRNA | BC137368.1 | - | 2 | 764 | 9021366 | |
BC137369.1 | - | 2 | 764 | 9021367 | ||
BC150189.1 | - | 2 | 776 | 40148511 | ||
BX647300.1 | - | 2 | 794 |
Gene Symbol: IFIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270927.1 | NP_001257856.1 | 3 | 978 | ||
NM_001270928.1 | NP_001257857.1 | 3 | 995 | |||
NM_001270929.1 | NP_001257858.1 | 3 | 990 | |||
NM_001270930.1 | NP_001257859.1 | 3 | 1116 | |||
NM_001548.4 | NP_001539.3 | 2 | 881 | |||
GenBank mRNA | AK092813.1 | - | 2 | 1909 | ||
AK308688.1 | - | 3 | 882 | |||
AK314588.1 | - | 2 | 804 | |||
AL548182.3 | - | 2 | 784 | |||
BC007091.1 | - | 2 | 806 | 4250452 | ||
BT006667.1 | - | 1 | 714 | |||
X03557.1 | - | 2 | 778 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
