siRNA Details
Gene Symbol: IGSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170961.1 | NP_001164432.1 | 18 | 3958 | ||
NM_001170962.1 | NP_001164433.1 | 17 | 3916 | |||
NM_001555.4 | NP_001546.2 | 18 | 3943 | |||
XM_011531330.1 | XP_011529632.1 | 19 | 3856 | |||
XM_011531333.1 | XP_011529635.1 | 18 | 3709 | |||
XM_011531334.1 | XP_011529636.1 | 16 | 3382 | |||
GenBank mRNA | AF034198.1 | - | 17 | 3765 | ||
AK226008.1 | - | 18 | 3815 | |||
AK295438.1 | - | 9 | 1994 | |||
AK307871.1 | - | 18 | 3819 | |||
AL137369.1 | - | 2 | 205 | |||
BC063884.1 | - | 18 | 3779 | 30387876 | ||
Y10523.1 | - | 17 | 3740 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
