siRNA Details
Gene Symbol: Mup4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008648.1 | NP_032674.1 | 3 | 311 | ||
GenBank mRNA | BC012227.1 | - | 3 | 298 | 4190774 | |
EU882229.1 | - | 3 | 245 | |||
M16358.1 | - | 3 | 311 |
Gene Symbol: Mup20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012323.1 | NP_001012323.1 | 3 | 322 | ||
GenBank mRNA | BC089613.1 | - | 3 | 322 | 6746470 | |
BC092096.1 | - | 3 | 348 | 4161847 | ||
EU882234.1 | - | 3 | 254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
