siRNA Details
Gene Symbol: Clec4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170332.1 | NP_001163803.1 | 2, 3 | 282 | ||
NM_001170333.1 | NP_001163804.1 | 1, 2 | 362 | |||
NM_011999.4 | NP_036129.1 | 1, 2 | 362 | |||
GenBank mRNA | AA387662.1 | - | 1, 2 | 368 | ||
AF387099.1 | - | 1, 2 | 367 | |||
AJ133533.1 | - | 1, 2 | 368 | |||
AK049002.1 | - | 1, 2 | 287 | |||
BC006623.1 | - | 1, 2 | 211 | 3968187 | ||
BC075729.1 | - | 1, 2 | 204 | 30287332 | ||
BC096565.1 | - | 2, 3 | 291 | 874396 |
Gene Symbol: Clec4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005860.2 | NP_001005860.1 | 1, 2 | 90 | ||
GenBank mRNA | AY397673.1 | - | 1, 2 | 90 | ||
BC104370.1 | - | 1, 2 | 90 | 40049888 | ||
BC104371.2 | - | 1, 2 | 90 |

More Information
Other Species Targeted

Gene Ontology Categories:
