siRNA Details
Gene Symbol: Tle4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF141905.1 | - | 1 | 1035 | ||
AK122481.2 | - | 18 | 3411 | |||
AK134989.1 | - | 20 | 4187 | |||
BC019750.1 | - | 4 | 1617 | |||
BC043477.1 | - | 5 | 1767 | |||
BC058525.1 | - | 3 | 1506 | |||
BC086631.1 | - | 21 | 4096 | |||
BC086780.1 | - | 3 | 1446 |

More Information
Other Species Targeted

