siRNA Details
Gene Symbol: Chchd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024166.6 | NP_077128.2 | 2 | 513 | ||
GenBank mRNA | AK003399.1 | - | 2 | 348 | ||
AK012691.1 | - | 1 | 339 | |||
AK088374.1 | - | 2 | 514 | |||
AK145492.1 | - | 2 | 331 | |||
AK160779.1 | - | 2 | 333 | |||
AK162133.1 | - | 2 | 331 | |||
AK166796.1 | - | 2 | 333 | |||
AK167593.1 | - | 2 | 333 | |||
BC037682.1 | - | 2 | 339 | |||
BC042717.1 | - | 2 | 320 | 3154544 | ||
BC082287.1 | - | 2 | 323 | 6397957 | ||
BC083126.1 | - | 2 | 302 | |||
BC084682.1 | - | 2 | 302 | 6530688 |
Gene Symbol: Gm6222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC158129.1 | - | 1 | 353 | 9007397 |
Gene Symbol: Zbed5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183088.2 | NP_898911.2 | 2 | 370 | ||
GenBank mRNA | AK010551.1 | - | 2 | 371 | ||
BC137610.1 | - | 2 | 368 | 8860629 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
