siRNA Details
Gene Symbol: Ypel3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025347.2 | NP_079623.1 | 4 | 452 | ||
NM_026875.2 | NP_081151.2 | 4 | 552 | |||
GenBank mRNA | AB098742.1 | - | 3 | 257 | ||
AF523353.1 | - | 4 | 442 | |||
AK002925.1 | - | 4 | 437 | |||
AK003371.1 | - | 4 | 453 | |||
AK004431.1 | - | 4 | 525 | |||
AK210119.1 | - | 3 | 232 | |||
BC009171.1 | - | 4 | 426 |

More Information
Other Species Targeted

Gene Ontology Categories:
