siRNA Details
Gene Symbol: Clec4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170332.1 | NP_001163803.1 | 3 | 289 | ||
NM_001170333.1 | NP_001163804.1 | 2 | 369 | |||
NM_011999.4 | NP_036129.1 | 2 | 369 | |||
GenBank mRNA | AA387662.1 | - | 2 | 375 | ||
AF387099.1 | - | 2 | 374 | |||
AJ133533.1 | - | 2 | 375 | |||
AK049002.1 | - | 2 | 294 | |||
BC006623.1 | - | 2 | 218 | 3968187 | ||
BC075729.1 | - | 2 | 211 | 30287332 | ||
BC096565.1 | - | 3 | 298 | 874396 |
Gene Symbol: Clec4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005860.2 | NP_001005860.1 | 2 | 97 | ||
GenBank mRNA | AY397673.1 | - | 2 | 97 | ||
BC104370.1 | - | 2 | 97 | 40049888 | ||
BC104371.2 | - | 2 | 97 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
