siRNA Details
Gene Symbol: ACAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130410.1 | NP_001123882.1 | 10 | 1467 | ||
NM_001607.3 | NP_001598.1 | 12 | 1746 | |||
NR_024024.1 | 10 | 1871 | ||||
XM_006713122.1 | XP_006713185.1 | 11 | 1451 | |||
XM_006713123.2 | XP_006713186.1 | 11 | 1588 | |||
XM_011533650.2 | XP_011531952.1 | 10 | 1614 | |||
GenBank mRNA | AF035295.1 | - | 10 | 1602 | ||
AK025138.1 | - | 12 | 1580 | |||
AK127051.1 | - | 6 | 4652 | |||
BC000635.2 | - | 12 | 1633 | 3345271 | ||
BC011977.1 | - | 12 | 1581 | 3860150 | ||
BC014474.1 | - | 10 | 1265 | 4908159 | ||
X14813.1 | - | 12 | 1575 | |||
X51460.1 | - | 10 | 1247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
