siRNA Details
Gene Symbol: Tle4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF141905.1 | - | 1 | 1045 | ||
AK122481.2 | - | 18 | 3421 | |||
AK134989.1 | - | 20 | 4197 | |||
BC019750.1 | - | 4 | 1627 | |||
BC043477.1 | - | 5 | 1777 | |||
BC058525.1 | - | 3 | 1516 | |||
BC086631.1 | - | 21 | 4106 | |||
BC086780.1 | - | 3 | 1456 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_019141, XM_006231128, XM_006231129, XM_008760286.
