siRNA Details
Gene Symbol: Olfr544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020289.2 | NP_064685.2 | 2 | 818 | ||
GenBank mRNA | AF121982.1 | - | 1 | 250 | ||
BC106830.2 | - | 1 | 642 | 40044631 | ||
BC126870.1 | - | 1 | 642 | 40044632 | ||
BC126871.1 | - | 1 | 642 | 40044633 |
