siRNA Details
Gene Symbol: Chchd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024166.6 | NP_077128.2 | 3 | 555 | ||
GenBank mRNA | AK003399.1 | - | 3 | 390 | ||
AK012691.1 | - | 2 | 381 | |||
AK088374.1 | - | 3 | 556 | |||
AK145492.1 | - | 3 | 373 | |||
AK160762.1 | - | 3 | 374 | |||
AK160779.1 | - | 3 | 375 | |||
AK162133.1 | - | 3 | 373 | |||
AK166796.1 | - | 3 | 375 | |||
AK167593.1 | - | 3 | 375 | |||
BC037682.1 | - | 2 | 1735 | |||
BC042717.1 | - | 3 | 362 | 3154544 | ||
BC069887.1 | - | 3 | 137 | |||
BC082287.1 | - | 3 | 365 | 6397957 | ||
BC083126.1 | - | 2 | 1698 | |||
BC084682.1 | - | 3 | 344 | 6530688 |
Gene Symbol: Gm6222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC158129.1 | - | 1 | 395 | 9007397 |
Gene Symbol: Zbed5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183088.2 | NP_898911.2 | 3 | 412 | ||
GenBank mRNA | AK010551.1 | - | 3 | 413 | ||
BC137610.1 | - | 3 | 410 | 8860629 |

More Information
Other Species Targeted

Gene Ontology Categories:
