siRNA Details
Gene Symbol: 1810037I17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024461.2 | NP_077781.1 | 2 | 303 | ||
GenBank mRNA | AK007721.1 | - | 2 | 304 | ||
AK009748.1 | - | 2 | 269 | |||
AK013091.1 | - | 2 | 288 | |||
AK170776.1 | - | 2 | 269 | |||
BC002135.1 | - | 2 | 312 | 3484100 | ||
BC086913.1 | - | 2 | 268 | 30121587 | ||
BC091739.1 | - | 2 | 271 |
