siRNA Details
Gene Symbol: 2310039H08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025966.3 | NP_080242.1 | 1 | 689 | ||
GenBank mRNA | AK003321.1 | - | 1 | 502 | ||
AK009696.1 | - | 1 | 512 | |||
BC038674.1 | - | 1 | 689 | 1513865 | ||
BU055991.1 | - | 1 | 484 |
